Assay Details
Gene Symbol: ARAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015230.3 | NP_056045.2 | 25 - 26 | 4373 | 136 | |
XM_017007698.1 | XP_016863187.1 | 25 - 26 | 4803 | 136 | ||
XM_017007699.1 | XP_016863188.1 | 25 - 26 | 4258 | 136 | ||
XM_017007700.1 | XP_016863189.1 | 24 - 25 | 4734 | 136 | ||
XM_017007701.1 | XP_016863190.1 | 25 - 26 | 4803 | 136 | ||
XM_017007702.1 | XP_016863191.1 | 24 - 25 | 4734 | 136 | ||
GenBank mRNA | AB011152.2 | - | 24 - 25 | 3816 | 136 | |
AF411982.1 | - | 14 - 15 | 1812 | 136 | ||
AF439781.1 | - | 24 - 25 | 4195 | 136 | ||
AY049733.1 | - | 25 - 26 | 4288 | 136 | ||
BC105089.1 | - | 12 - 13 | 1587 | 136 | ||
BC105091.1 | - | 12 - 13 | 1587 | 136 | ||
BC150258.1 | - | 24 - 25 | 3827 | 136 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04002955.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
