Assay Details
Gene Symbol: SPHKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142644.1 | NP_001136116.1 | 7 - 8 | 4493 | 60 | |
NM_030623.3 | NP_085126.2 | 7 - 8 | 4493 | 60 | ||
XM_005246870.4 | XP_005246927.1 | 8 - 9 | 4965 | 60 | ||
XM_006712777.3 | XP_006712840.1 | 8 - 9 | 4926 | 60 | ||
XM_006712778.3 | XP_006712841.1 | 8 - 9 | 4925 | 60 | ||
XM_011511925.2 | XP_011510227.1 | 9 - 10 | 5044 | 60 | ||
GenBank mRNA | AB051465.1 | - | 1 - 2 | 3341 | 60 | |
AK090876.1 | - | 1 - 2 | 1568 | 60 | ||
AK095042.1 | - | 1 - 2 | 655 | 60 | ||
BC104954.1 | - | 7 - 8 | 4493 | 8143957 | 60 | |
BC104956.1 | - | 7 - 8 | 4493 | 8143959 | 60 | |
CR749297.1 | - | 1 - 2 | 3599 | 60 | ||
CR749494.1 | - | 7 - 8 | 4475 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
