Assay Details
Gene Symbol: UGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003358.2 | NP_003349.1 | 7 - 8 | 1273 | 124 | |
XM_005252186.4 | XP_005252243.1 | 8 - 9 | 1296 | 124 | ||
GenBank mRNA | AK314847.1 | - | 7 - 8 | 878 | 124 | |
BC038711.1 | - | 7 - 8 | 1274 | 5295561 | 124 | |
D50840.1 | - | 7 - 8 | 1113 | 124 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000560.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09006259.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09006259.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09006259.hg.1 | TC09000560.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156438.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156438.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156438.hg.1 | TC0900008441.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900156438.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156438.hg.1 | ENST00000374279.3 | Ensembl |
