Assay Details
Gene Symbol: ABCA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015657.3 | NP_056472.2 | 35 - 36 | 5599 | 90 | |
NM_173076.2 | NP_775099.2 | 43 - 44 | 6614 | 90 | ||
NR_103740.1 | 45 - 46 | 6694 | 90 | |||
XM_011510951.2 | XP_011509253.1 | 43 - 44 | 6786 | 90 | ||
GenBank mRNA | AF418105.1 | - | 38 - 39 | 5736 | 90 | |
AK123578.1 | - | 10 - 11 | 1154 | 90 | ||
AY033486.1 | - | 35 - 36 | 5599 | 90 | ||
AY219711.1 | - | 43 - 44 | 6614 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002746.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
