Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000487237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000487237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
ENST00000487237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036170.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036170.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036170.hg.1 |
TC01002345.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100198559.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198559.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198559.hg.1 |
TC0100013287.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198559.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198559.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000361729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000487237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000487237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
ENST00000487237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013287.hg.1 |
NM_004091.3 |
Refseq |