Assay Details
Gene Symbol: BNIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159642.1 | NP_001153114.1 | 6 - 7 | 1148 | 98 | |
NM_138278.3 | NP_612122.2 | 6 - 7 | 896 | 98 | ||
XM_011509236.1 | XP_011507538.1 | 6 - 7 | 891 | 98 | ||
XM_017000420.1 | XP_016855909.1 | 7 - 8 | 943 | 98 | ||
GenBank mRNA | AF193056.1 | - | 6 - 7 | 1094 | 98 | |
AK092595.1 | - | 6 - 7 | 848 | 98 | ||
AY033000.1 | - | 6 - 7 | 880 | 98 | ||
AY078983.1 | - | 3 - 4 | 477 | 98 | ||
AY078984.1 | - | 3 - 4 | 477 | 98 | ||
BC027868.1 | - | 6 - 7 | 873 | 5205026 | 98 | |
BC074779.2 | - | 3 - 4 | 518 | 30915557 | 98 | |
BC074780.2 | - | 3 - 4 | 518 | 30915335 | 98 | |
CU451236.1 | - | 2 - 3 | 238 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001199.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01018503.hg.1 | ENST00000295294.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018503.hg.1 | ENST00000361277.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018503.hg.1 | ENST00000368931.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018503.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018503.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018503.hg.1 | TC01001199.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | TC0100009936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171867.hg.1 | NM_138278.3 | Refseq |
