Assay Details
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148894.2 | NP_683692.2 | 18 - 19 | 8640 | 76 | |
XM_005248150.3 | XP_005248207.1 | 18 - 19 | 8657 | 76 | ||
XM_005248151.3 | XP_005248208.1 | 18 - 19 | 8657 | 76 | ||
XM_006713958.3 | XP_006714021.1 | 18 - 19 | 8657 | 76 | ||
XM_011513827.2 | XP_011512129.1 | 18 - 19 | 8657 | 76 | ||
XM_011513829.2 | XP_011512131.1 | 18 - 19 | 8657 | 76 | ||
XM_011513830.2 | XP_011512132.1 | 16 - 17 | 8040 | 76 | ||
XM_017008009.1 | XP_016863498.1 | 18 - 19 | 8657 | 76 | ||
XM_017008010.1 | XP_016863499.1 | 18 - 19 | 8657 | 76 | ||
GenBank mRNA | AB037748.2 | - | 9 - 10 | 4784 | 76 | |
AK025361.1 | - | 4 - 5 | 301 | 76 | ||
AK122806.1 | - | 6 - 7 | 1973 | 76 | ||
AL137350.1 | - | 9 - 10 | 1195 | 76 | ||
BC016987.1 | - | 8 - 9 | 580 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001047.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
