Assay Details
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148894.2 | NP_683692.2 | 22 - 23 | 8862 | 155 | |
XM_005248150.3 | XP_005248207.1 | 22 - 23 | 8879 | 155 | ||
XM_005248151.3 | XP_005248208.1 | 22 - 23 | 8876 | 155 | ||
XM_017008010.1 | XP_016863499.1 | 22 - 23 | 8876 | 155 | ||
GenBank mRNA | AB037748.2 | - | 13 - 14 | 5006 | 155 | |
AK025361.1 | - | 8 - 9 | 523 | 155 | ||
BC016987.1 | - | 12 - 13 | 802 | 155 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001047.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
