Assay Details
Gene Symbol: AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302959.1 | NP_001289888.1 | ||||
NM_001302960.1 | NP_001289889.1 | |||||
NM_003977.3 | NP_003968.3 | |||||
GenBank mRNA | BC104797.1 | - | 8143800 | |||
BC104827.1 | - | 8143830 | ||||
BG821308.1 | - | |||||
BQ054206.1 | - | |||||
BQ420398.1 | - | |||||
BU689235.1 | - | |||||
U31913.1 | - | |||||
U78521.1 | - |

More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to mitochondrion
xenobiotic metabolic process
signal transduction
regulation of protein kinase A signaling
protein maturation by protein folding
negative regulation of cyclic-nucleotide phosphodiesterase activity
regulation of nucleic acid-templated transcription
transcription coactivator activity
protein binding
transcription factor binding
aryl hydrocarbon receptor binding
GAF domain binding
unfolded protein binding
protein targeting to mitochondrion
xenobiotic metabolic process
signal transduction
regulation of protein kinase A signaling
protein maturation by protein folding
negative regulation of cyclic-nucleotide phosphodiesterase activity
regulation of nucleic acid-templated transcription
transcription coactivator activity
protein binding
transcription factor binding
aryl hydrocarbon receptor binding
GAF domain binding
unfolded protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000279146.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000525341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000528641.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | ENST00000529797.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302959.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_001302960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008139.hg.1 | NM_003977.3 | Refseq |
