Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017045.hg.1 |
TC19001212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017045.hg.1 |
ENST00000222219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017045.hg.1 |
ENST00000538460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017045.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017043.hg.1 |
TC19001212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017043.hg.1 |
ENST00000222219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017043.hg.1 |
ENST00000538460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017043.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000586547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000586547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000586547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017044.hg.1 |
TC19001212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017044.hg.1 |
ENST00000222219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017044.hg.1 |
ENST00000538460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017044.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007102.hg.1 |
ENST00000592400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007102.hg.1 |
ENST00000592400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017047.hg.1 |
TC19001212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017047.hg.1 |
ENST00000222219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017047.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179669.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179669.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179669.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179669.hg.1 |
TC1900009754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179669.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179669.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179669.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179674.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179674.hg.1 |
TC1900009754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179674.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179670.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179670.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179670.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179670.hg.1 |
TC1900009754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179670.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179670.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179670.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000586547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000586547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000586547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000588777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009754.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179672.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179672.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179672.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900179672.hg.1 |
TC1900009754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900179672.hg.1 |
ENST00000222219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179672.hg.1 |
ENST00000592506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900179672.hg.1 |
NM_001375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007102.hg.1 |
ENST00000592400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007102.hg.1 |
ENST00000592400.1 |
Ensembl |