Assay Details
Gene Symbol: GLRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042543.2 | NP_001036008.1 | 7 - 8 | 1427 | 105 | |
NM_006529.3 | NP_006520.2 | 7 - 8 | 1427 | 105 | ||
XM_011532268.2 | XP_011530570.1 | 3 - 4 | 673 | 105 | ||
XM_017008630.1 | XP_016864119.1 | 8 - 9 | 1451 | 105 | ||
GenBank mRNA | AF018157.1 | - | 1 - 2 | 190 | 105 | |
BC036086.1 | - | 7 - 8 | 1426 | 5286731 | 105 | |
U93917.1 | - | 7 - 8 | 1343 | 105 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001755.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neuropeptide signaling pathway
ion transmembrane transport
response to amino acid
protein homooligomerization
synaptic transmission, glycinergic
chloride transmembrane transport
glycine binding
extracellular-glycine-gated chloride channel activity
glycine-gated chloride ion channel activity
metal ion binding
neuropeptide signaling pathway
ion transmembrane transport
response to amino acid
protein homooligomerization
synaptic transmission, glycinergic
chloride transmembrane transport
glycine binding
extracellular-glycine-gated chloride channel activity
glycine-gated chloride ion channel activity
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04024202.hg.1 | TC04001755.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04024202.hg.1 | ENST00000274093.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024202.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024202.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024202.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | ENST00000274093.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | ENST00000340217.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | NM_001042543.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | NM_006529.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185447.hg.1 | TC0400012462.hg.1 | Clariom D TC |
