Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
ENST00000354454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
ENST00000536173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
ENST00000547044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
ENST00000548766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
NM_001204126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
NM_001204127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
NM_006152.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003045.hg.1 |
TC12000251.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000354454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000361433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000536173.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000547044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000548766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000549437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000555194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000555877.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
NM_001204126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
NM_001204127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
NM_006152.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000354454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000361433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000536173.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000547044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000548766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000549437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000555194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
ENST00000555877.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
NM_001204126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
NM_001204127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
NM_006152.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199537.hg.1 |
TC1200012606.hg.1 |
Clariom D TC |