Assay Details
Gene Symbol: MPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000250.1 | NP_000241.1 | 8 - 9 | 1537 | 102 | |
GenBank mRNA | BC130476.1 | - | 8 - 9 | 1411 | 40146507 | 102 |
J02694.1 | - | 8 - 9 | 1508 | 102 | ||
M19507.1 | - | 8 - 9 | 1537 | 102 | ||
S56200.1 | - | 8 - 9 | 1360 | 102 | ||
X04876.1 | - | 8 - 9 | 1523 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to yeast
hypochlorous acid biosynthetic process
respiratory burst involved in defense response
defense response
response to oxidative stress
aging
response to mechanical stimulus
removal of superoxide radicals
response to food
response to lipopolysaccharide
low-density lipoprotein particle remodeling
hydrogen peroxide catabolic process
negative regulation of apoptotic process
negative regulation of growth of symbiont in host
defense response to fungus
oxidation-reduction process
response to gold nanoparticle
chromatin binding
peroxidase activity
heparin binding
heme binding
metal ion binding
response to yeast
hypochlorous acid biosynthetic process
respiratory burst involved in defense response
defense response
response to oxidative stress
aging
response to mechanical stimulus
removal of superoxide radicals
response to food
response to lipopolysaccharide
low-density lipoprotein particle remodeling
hydrogen peroxide catabolic process
negative regulation of apoptotic process
negative regulation of growth of symbiont in host
defense response to fungus
oxidation-reduction process
response to gold nanoparticle
chromatin binding
peroxidase activity
heparin binding
heme binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000577220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000577220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000577220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000581022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000581022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | ENST00000581022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700190419.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700190419.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700190419.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700190419.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700190419.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700190419.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700190419.hg.1 | TC1700011249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700190418.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700190418.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700190418.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700190418.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700190418.hg.1 | TC1700011249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000225275.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000577220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000577220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000577220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000578493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000580005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000581022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000581022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | ENST00000581022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011249.hg.1 | NM_000250.1 | Refseq |
