Assay Details
Gene Symbol: CERK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022766.5 | NP_073603.2 | 11 - 12 | 1439 | 94 | |
XM_017028909.1 | XP_016884398.1 | 11 - 12 | 1236 | 94 | ||
GenBank mRNA | AB051433.1 | - | 10 - 11 | 1165 | 94 | |
AB079066.1 | - | 11 - 12 | 1455 | 94 | ||
AJ457828.1 | - | 11 - 12 | 1402 | 94 | ||
AK291476.1 | - | 11 - 12 | 1395 | 94 | ||
BC067255.1 | - | 10 - 11 | 1316 | 94 | ||
BC126940.1 | - | 11 - 12 | 1382 | 40084110 | 94 | |
CR456404.1 | - | 11 - 12 | 1342 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000890.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22014971.hg.1 | TC22000890.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22014971.hg.1 | ENST00000216264.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014971.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014971.hg.1 | ENST00000541677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014971.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | TC2200009028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165410.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
