Assay Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020945.1 | NP_065996.1 | 43 - 44 | 7246 | 87 | |
XM_005270004.3 | XP_005270061.1 | 43 - 44 | 7361 | 87 | ||
XM_011539986.2 | XP_011538288.2 | 43 - 44 | 7364 | 87 | ||
XM_011539987.2 | XP_011538289.2 | 43 - 44 | 7367 | 87 | ||
XM_011539988.2 | XP_011538290.1 | 44 - 45 | 7445 | 87 | ||
XM_017016463.1 | XP_016871952.1 | 44 - 45 | 7288 | 87 | ||
XM_017016464.1 | XP_016871953.1 | 25 - 26 | 3675 | 87 | ||
GenBank mRNA | AB046827.1 | - | 10 - 11 | 1429 | 87 | |
AK074085.1 | - | 23 - 24 | 3304 | 87 | ||
AK074443.1 | - | 5 - 6 | 649 | 87 | ||
AL833669.1 | - | 3 - 4 | 375 | 87 | ||
BC047574.1 | - | 3 - 4 | 315 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000323.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
