Assay Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020945.1 | NP_065996.1 | 53 - 54 | 8304 | 69 | |
XM_005270004.3 | XP_005270061.1 | 53 - 54 | 8419 | 69 | ||
XM_011539986.2 | XP_011538288.2 | 53 - 54 | 8422 | 69 | ||
XM_011539987.2 | XP_011538289.2 | 53 - 54 | 8425 | 69 | ||
XM_011539988.2 | XP_011538290.1 | 54 - 55 | 8503 | 69 | ||
XM_017016463.1 | XP_016871952.1 | 54 - 55 | 8346 | 69 | ||
XM_017016464.1 | XP_016871953.1 | 35 - 36 | 4733 | 69 | ||
GenBank mRNA | AB046827.1 | - | 21 - 22 | 2487 | 69 | |
AK024502.1 | - | 4 - 5 | 471 | 69 | ||
AK074085.1 | - | 34 - 35 | 4362 | 69 | ||
AK074443.1 | - | 16 - 17 | 1707 | 69 | ||
AK301736.1 | - | 7 - 8 | 760 | 69 | ||
AK310374.1 | - | 7 - 8 | 804 | 69 | ||
AL833669.1 | - | 14 - 15 | 1433 | 69 | ||
BC047574.1 | - | 14 - 15 | 1373 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000323.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
