Assay Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020945.1 | NP_065996.1 | 31 - 32 | 5412 | 65 | |
XM_005270004.3 | XP_005270061.1 | 31 - 32 | 5527 | 65 | ||
XM_011539986.2 | XP_011538288.2 | 31 - 32 | 5530 | 65 | ||
XM_011539987.2 | XP_011538289.2 | 31 - 32 | 5533 | 65 | ||
XM_011539988.2 | XP_011538290.1 | 32 - 33 | 5611 | 65 | ||
XM_011539990.2 | XP_011538292.1 | 31 - 32 | 5558 | 65 | ||
XM_011539991.2 | XP_011538293.1 | 31 - 32 | 5562 | 65 | ||
XM_017016463.1 | XP_016871952.1 | 32 - 33 | 5454 | 65 | ||
XM_017016464.1 | XP_016871953.1 | 13 - 14 | 1841 | 65 | ||
GenBank mRNA | AK074085.1 | - | 11 - 12 | 1470 | 65 | |
AK093607.1 | - | 8 - 9 | 974 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000323.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
