Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008886.hg.1 |
TC05000641.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008886.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008886.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159311.hg.1 |
TC0500008644.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008644.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
ENST00000495905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159314.hg.1 |
TC0500008644.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
ENST00000231449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
ENST00000350025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
ENST00000622422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
NM_000589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
NM_172348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500159312.hg.1 |
TC0500008644.hg.1 |
Clariom D TC |