Assay Details
Gene Symbol: A2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000014.4 | NP_000005.2 | 35 - 36 | 4521 | 80 | |
GenBank mRNA | AB209614.2 | - | 35 - 36 | 4432 | 80 | |
AK307832.1 | - | 34 - 35 | 4258 | 80 | ||
BC026246.1 | - | 17 - 18 | 2149 | 80 | ||
BC040071.1 | - | 35 - 36 | 4460 | 6056126 | 80 | |
BX647329.1 | - | 18 - 19 | 2198 | 80 | ||
BX648622.1 | - | 3905 | 80 | |||
M36501.1 | - | 16 - 17 | 1915 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001187.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
