Assay Details
Gene Symbol: A2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000014.4 | NP_000005.2 | 4 - 5 | 588 | 84 | |
XM_006719056.2 | XP_006719119.1 | 4 - 5 | 566 | 84 | ||
GenBank mRNA | AB209614.2 | - | 4 - 5 | 499 | 84 | |
AK307832.1 | - | 3 - 4 | 325 | 84 | ||
AK312482.1 | - | 4 - 5 | 489 | 84 | ||
AY591530.1 | - | 4 - 5 | 553 | 84 | ||
BC026246.1 | - | 4 - 5 | 544 | 84 | ||
BC040071.1 | - | 4 - 5 | 527 | 6056126 | 84 | |
CR749334.1 | - | 4 - 5 | 546 | 84 | ||
M11313.1 | - | 4 - 5 | 518 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
