Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
TC0X001427.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000338585.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000370573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000370576.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000370578.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000414978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000519895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000520602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
ENST00000536274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
NM_001099855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
NM_001171876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
NM_001171877.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
NM_001171878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
NM_001171879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018930.hg.1 |
NM_005369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000338585.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000370573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000370576.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000414978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000437564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000446225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000519895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000520602.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000536274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001099855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_005369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000338585.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000370573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000370576.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000414978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000437564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000446225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000519895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000520602.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
ENST00000536274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001099855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_001171879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
NM_005369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174497.hg.1 |
TC0X00010962.hg.1 |
Clariom D TC |