Assay Details
Gene Symbol: PBXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317734.1 | NP_001304663.1 | 3 - 4 | 288 | 64 | |
NM_020524.3 | NP_065385.2 | 4 - 5 | 377 | 64 | ||
GenBank mRNA | AF221521.1 | - | 4 - 5 | 326 | 64 | |
AK022497.1 | - | 3 - 4 | 247 | 64 | ||
AK023219.1 | - | 4 - 5 | 318 | 64 | ||
AK300544.1 | - | 4 - 5 | 341 | 64 | ||
AK310388.1 | - | 5 - 6 | 806 | 64 | ||
AK315551.1 | - | 4 - 5 | 358 | 64 | ||
BC016852.1 | - | 4 - 5 | 305 | 3900461 | 64 | |
HY108652.1 | - | 4 - 5 | 377 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
