Assay Details
Gene Symbol: PBXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317734.1 | NP_001304663.1 | 5 - 6 | 692 | 76 | |
NM_001317735.1 | NP_001304664.1 | 3 - 4 | 361 | 76 | ||
NM_020524.3 | NP_065385.2 | 6 - 7 | 781 | 76 | ||
GenBank mRNA | AF221521.1 | - | 6 - 7 | 730 | 76 | |
AK022497.1 | - | 5 - 6 | 651 | 76 | ||
AK023219.1 | - | 6 - 7 | 722 | 76 | ||
AK303413.1 | - | 3 - 4 | 305 | 76 | ||
AK303863.1 | - | 3 - 4 | 206 | 76 | ||
AK315551.1 | - | 6 - 7 | 762 | 76 | ||
BC016852.1 | - | 6 - 7 | 709 | 3900461 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003298.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000351146.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000368460.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000368463.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000368465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000498553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000539880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000542459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | ENST00000543593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01050911.hg.1 | TC01003298.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | TC0100015932.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000368460.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219966.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | NM_020524.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | TC0100015932.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000368463.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000368465.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000490230.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | ENST00000498553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100219968.hg.1 | NM_020524.3 | Refseq |
