Assay Details
Gene Symbol: TESK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006285.2 | NP_006276.2 | 5 - 6 | 956 | 58 | |
GenBank mRNA | AF507907.1 | - | 4 - 5 | 369 | 58 | |
AK298907.1 | - | 5 - 6 | 494 | 58 | ||
BC037561.1 | - | 5 - 6 | 551 | 58 | ||
BC038448.1 | - | 5 - 6 | 930 | 5170888 | 58 | |
BC067130.1 | - | 5 - 6 | 967 | 6153652 | 58 | |
D50863.1 | - | 5 - 6 | 892 | 58 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000187.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09002085.hg.1 | TC09000187.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09002085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002085.hg.1 | ENST00000498522.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002085.hg.1 | ENST00000535770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NR_039813.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148136.hg.1 | TC0900007085.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NR_039813.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 142009 | - | - |
