Assay Details
Gene Symbol: TESK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318230.1 | NP_001305159.1 | 5 - 6 | 964 | 85 | |
NM_006285.2 | NP_006276.2 | 6 - 7 | 1047 | 85 | ||
GenBank mRNA | AF507907.1 | - | 5 - 6 | 460 | 85 | |
AK298907.1 | - | 6 - 7 | 585 | 85 | ||
BC037561.1 | - | 6 - 7 | 642 | 85 | ||
BC038448.1 | - | 6 - 7 | 1021 | 5170888 | 85 | |
BC067130.1 | - | 6 - 7 | 1058 | 6153652 | 85 | |
D50863.1 | - | 6 - 7 | 983 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000187.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09002088.hg.1 | TC09000187.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09002088.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002088.hg.1 | ENST00000498522.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002088.hg.1 | ENST00000535770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002088.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NR_039813.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148139.hg.1 | TC0900007085.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NR_039813.2 | Refseq |
