Assay Details
Gene Symbol: AKAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005858.3 | NP_005849.1 | 11 - 12 | 1473 | 70 | |
XM_011527624.2 | XP_011525926.1 | 11 - 12 | 1566 | 70 | ||
XM_017026141.1 | XP_016881630.1 | 11 - 12 | 1618 | 70 | ||
GenBank mRNA | AK309529.1 | - | 8 - 9 | 1001 | 70 | |
AL050160.1 | - | 5 - 6 | 401 | 70 | ||
BC037270.2 | - | 11 - 12 | 1463 | 4837309 | 70 | |
Y11997.1 | - | 11 - 12 | 1464 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001253.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
RNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
RNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19017617.hg.1 | TC19001253.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19017617.hg.1 | ENST00000269701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017617.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180872.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180872.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180872.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180872.hg.1 | TC1900009866.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900180872.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180872.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180872.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
