Assay Details
Gene Symbol: AKAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005858.3 | NP_005849.1 | 6 - 7 | 1066 | 67 | |
XM_011527624.2 | XP_011525926.1 | 6 - 7 | 1159 | 67 | ||
XM_017026141.1 | XP_016881630.1 | 6 - 7 | 1211 | 67 | ||
GenBank mRNA | AK303640.1 | - | 3 - 4 | 693 | 67 | |
AK308251.1 | - | 4 - 5 | 802 | 67 | ||
AK309529.1 | - | 2 - 3 | 436 | 67 | ||
BC037270.2 | - | 6 - 7 | 1056 | 4837309 | 67 | |
Y11997.1 | - | 6 - 7 | 1057 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001253.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19017624.hg.1 | ENST00000269701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017624.hg.1 | ENST00000537303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017624.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19017624.hg.1 | TC19001253.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19017621.hg.1 | ENST00000269701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017621.hg.1 | ENST00000537303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017621.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19017621.hg.1 | TC19001253.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19017623.hg.1 | ENST00000269701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017623.hg.1 | ENST00000537303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017623.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19017623.hg.1 | TC19001253.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | TC1900009866.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180880.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | TC1900009866.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | ENST00000599883.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180879.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | NM_005858.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | TC1900009866.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | ENST00000269701.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | ENST00000537303.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | ENST00000598597.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180876.hg.1 | NM_005858.3 | Refseq |
