Assay Details
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206802.2 | NP_001193731.1 | 2 - 3 | 409 | 69 | |
NM_004881.4 | NP_004872.2 | 2 - 3 | 1293 | 69 | ||
NM_147184.3 | NP_671713.1 | 3 - 4 | 894 | 69 | ||
XM_006712150.2 | XP_006712213.1 | 3 - 4 | 894 | 69 | ||
GenBank mRNA | AF010309.1 | - | 2 - 3 | 931 | 69 | |
AK223382.1 | - | 2 - 3 | 1130 | 69 | ||
AY371700.1 | - | 2 - 3 | 404 | 69 | ||
BC000474.2 | - | 3 - 4 | 464 | 2961571 | 69 | |
BC018819.2 | - | 3 - 4 | 464 | 69 | ||
BT007149.1 | - | 2 - 3 | 404 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001645.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000470636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000470636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | ENST00000470636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | TC02001645.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | ENST00000238721.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | ENST00000313482.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | ENST00000335934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | ENST00000407482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026229.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | TC02001645.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | ENST00000238721.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | ENST00000313482.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | ENST00000335934.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | ENST00000407482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026230.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186996.hg.1 | TC0200011975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200186997.hg.1 | TC0200011975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000238721.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000335934.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000407482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000413037.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000417886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000470636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000470636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | ENST00000470636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_001206802.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_004881.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011975.hg.1 | NM_147184.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 215041 | - | - |
