Assay Details
Gene Symbol: CAND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162499.1 | NP_001155971.1 | 13 - 14 | 3425 | 57 | |
NM_012298.2 | NP_036430.1 | 11 - 12 | 3074 | 57 | ||
XM_011533504.2 | XP_011531806.1 | 13 - 14 | 3539 | 57 | ||
GenBank mRNA | AB014567.1 | - | 11 - 12 | 3002 | 57 | |
BC136592.1 | - | 13 - 14 | 3364 | 9020582 | 57 | |
BX648800.1 | - | 4 - 5 | 578 | 57 | ||
GU727621.1 | - | 13 - 14 | 3425 | 57 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000074.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03001646.hg.1 | ENST00000295989.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03001646.hg.1 | ENST00000454887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03001646.hg.1 | ENST00000456430.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03001646.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03001646.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03001646.hg.1 | TC03000074.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000626378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000626378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | ENST00000626378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000446928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000466558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000626378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000626378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | ENST00000626378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013789.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | NM_001162499.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | TC0300013789.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | NM_012298.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | ENST00000295989.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | ENST00000456430.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203444.hg.1 | NM_001162499.1 | Refseq |
