Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
ENST00000286437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
ENST00000342251.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
ENST00000370457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
ENST00000370460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
NM_001169122.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
NM_001169123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
NM_001169124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
NM_001169125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
NM_001170628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
NM_002025.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009087.hg.1 |
TC0X000708.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000286437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000342251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000370457.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000370460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169122.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001170628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_002025.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
TC0X00008674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000286437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000342251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000370457.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
ENST00000370460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169122.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001169125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_001170628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158786.hg.1 |
NM_002025.3 |
Refseq |