Assay Details
Gene Symbol: AFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169123.1 | NP_001162594.1 | 4 - 5 | 1554 | 62 | |
NM_002025.3 | NP_002016.2 | 4 - 5 | 1566 | 62 | ||
GenBank mRNA | BC132683.1 | - | 4 - 5 | 1126 | 40146705 | 62 |
U48436.1 | - | 4 - 5 | 1566 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000708.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
