Assay Details
Gene Symbol: YEATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018023.4 | NP_060493.3 | 12 - 13 | 1742 | 116 | |
XM_005247597.2 | XP_005247654.1 | 12 - 13 | 1759 | 116 | ||
XM_005247598.4 | XP_005247655.1 | 12 - 13 | 1667 | 116 | ||
XM_006713688.3 | XP_006713751.1 | 12 - 13 | 1877 | 116 | ||
XM_011512965.1 | XP_011511267.1 | 11 - 12 | 1597 | 116 | ||
XM_011512966.1 | XP_011511268.1 | 12 - 13 | 1759 | 116 | ||
XM_017006809.1 | XP_016862298.1 | 12 - 13 | 1873 | 116 | ||
XM_017006810.1 | XP_016862299.1 | 12 - 13 | 1664 | 116 | ||
XM_017006811.1 | XP_016862300.1 | 11 - 12 | 1529 | 116 | ||
XM_017006812.1 | XP_016862301.1 | 12 - 13 | 1763 | 116 | ||
XM_017006813.1 | XP_016862302.1 | 11 - 12 | 1563 | 116 | ||
XM_017006814.1 | XP_016862303.1 | 11 - 12 | 1544 | 116 | ||
GenBank mRNA | AB033023.1 | - | 12 - 13 | 1742 | 116 | |
BC150273.1 | - | 12 - 13 | 1742 | 9007245 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000986.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03016965.hg.1 | ENST00000305135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03016965.hg.1 | ENST00000421660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03016965.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03016965.hg.1 | TC03000986.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300169721.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300169721.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300169721.hg.1 | TC0300009677.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300169721.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300169721.hg.1 | ENST00000305135.9 | Ensembl |
