Assay Details
Gene Symbol: KIF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004523.3 | NP_004514.2 | 14 - 15 | 2186 | 157 | |
GenBank mRNA | AK314258.1 | - | 14 - 15 | 2089 | 157 | |
BC126211.1 | - | 14 - 15 | 1985 | 8991927 | 157 | |
BC136474.1 | - | 14 - 15 | 1985 | 9020463 | 157 | |
U37426.1 | - | 14 - 15 | 1968 | 157 | ||
X85137.1 | - | 14 - 15 | 1888 | 157 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000659.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
retrograde vesicle-mediated transport, Golgi to endoplasmic reticulum
microtubule-based movement
spindle organization
mitotic spindle organization
chromosome segregation
mitotic centrosome separation
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of mitotic centrosome separation
cell division
mitotic spindle assembly
microtubule motor activity
ATP binding
microtubule binding
plus-end-directed microtubule motor activity
protein kinase binding
protein-containing complex binding
mitotic cell cycle
retrograde vesicle-mediated transport, Golgi to endoplasmic reticulum
microtubule-based movement
spindle organization
mitotic spindle organization
chromosome segregation
mitotic centrosome separation
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of mitotic centrosome separation
cell division
mitotic spindle assembly
microtubule motor activity
ATP binding
microtubule binding
plus-end-directed microtubule motor activity
protein kinase binding
protein-containing complex binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10007386.hg.1 | ENST00000260731.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10007386.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10007386.hg.1 | TC10000659.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000157526.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000157526.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000157526.hg.1 | TC1000008471.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000157526.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000157526.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
