Assay Details
Gene Symbol: TTC41P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027249.1 | 11 - 12 | 3271 | 61 | ||
GenBank mRNA | BC064342.1 | - | 1 - 2 | 161 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | 5'-nucleotidase domain containing 3 | NT5DC3 |
TU12B1-TY, TU12B1TY |
-20.9kb | Sense | Chr. 12 - 103746315 - 103841238 on NCBI Build GRCh38 |
Downstream | heat shock protein 90 beta family member 1 | HSP90B1 |
ECGP, GP96, GRP94, HEL-S-125m, HEL35, TRA1 |
63.8kb | Anti-sense | Chr. 12 - 103930334 - 103947930 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001902.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12024806.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12024806.hg.1 | TC12001902.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12024804.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12024804.hg.1 | TC12001902.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12024805.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12024805.hg.1 | TC12001902.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12024807.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12024807.hg.1 | TC12001902.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191068.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191068.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191068.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191068.hg.1 | TC1200011749.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200191068.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191068.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191068.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191067.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191067.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191067.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191067.hg.1 | TC1200011749.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200191067.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191067.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191067.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191069.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191069.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191069.hg.1 | TC1200011749.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200191069.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191069.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191065.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191065.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191065.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191065.hg.1 | TC1200011749.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200191065.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191065.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191065.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191066.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191066.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191066.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191066.hg.1 | TC1200011749.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200191066.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191066.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191066.hg.1 | NR_027249.1 | Refseq |
