Assay Details
Gene Symbol: LOC101929715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AA631078.1 | - | 1 - 2 | 317 | 78 | |
AW303360.1 | - | 2 - 3 | 154 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
