Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000565038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000565038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000565038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013914.hg.1 |
ENST00000261584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013914.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013914.hg.1 |
TC16000967.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600175110.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175110.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175110.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175110.hg.1 |
TC1600009746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600175110.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175110.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175110.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000261584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000561514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000565038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000565038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000565038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000566069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000567003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
ENST00000568219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009746.hg.1 |
NM_024675.3 |
Refseq |