Assay Details
Gene Symbol: ERBB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042599.1 | NP_001036064.1 | 24 - 25 | 3065 | 64 | |
NM_005235.2 | NP_005226.1 | 24 - 25 | 3065 | 64 | ||
XM_005246376.2 | XP_005246433.1 | 24 - 25 | 3554 | 64 | ||
XM_005246377.2 | XP_005246434.1 | 24 - 25 | 3554 | 64 | ||
XM_006712364.2 | XP_006712427.1 | 25 - 26 | 3629 | 64 | ||
XM_017003577.1 | XP_016859066.1 | 26 - 27 | 3707 | 64 | ||
XM_017003578.1 | XP_016859067.1 | 25 - 26 | 3662 | 64 | ||
XM_017003579.1 | XP_016859068.1 | 26 - 27 | 3707 | 64 | ||
XM_017003580.1 | XP_016859069.1 | 25 - 26 | 3632 | 64 | ||
XM_017003581.1 | XP_016859070.1 | 25 - 26 | 3662 | 64 | ||
XM_017003582.1 | XP_016859071.1 | 21 - 22 | 2742 | 64 | ||
GenBank mRNA | AB209697.1 | - | 14 - 15 | 1769 | 64 | |
BC112199.1 | - | 24 - 25 | 3006 | 8327667 | 64 | |
BC143741.1 | - | 24 - 25 | 2994 | 9052255 | 64 | |
BC143747.1 | - | 24 - 25 | 2964 | 9052261 | 64 | |
BC143749.1 | - | 24 - 25 | 2964 | 9052263 | 64 | |
KT281867.1 | - | 5 - 6 | 872 | 64 | ||
KT310076.1 | - | 13 - 14 | 1752 | 64 | ||
L07868.1 | - | 24 - 25 | 3000 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002735.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
