Assay Details
Gene Symbol: CAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018448.4 | NP_060918.2 | 5 - 6 | 1181 | 150 | |
GenBank mRNA | AB020636.2 | - | 5 - 6 | 1083 | 150 | |
AK074922.1 | - | 2 - 3 | 355 | 150 | ||
AK096010.1 | - | 5 - 6 | 1087 | 150 | ||
AK292456.1 | - | 7 - 8 | 1020 | 150 | ||
AK314358.1 | - | 5 - 6 | 1069 | 150 | ||
AL136810.1 | - | 5 - 6 | 1093 | 150 | ||
BC026220.2 | - | 3 - 4 | 532 | 150 | ||
BC050341.1 | - | 5 - 6 | 877 | 5265409 | 150 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000595.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12007841.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007841.hg.1 | ENST00000540047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007841.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007841.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007841.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007842.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007842.hg.1 | ENST00000540047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007842.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007842.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007842.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007843.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007843.hg.1 | ENST00000540047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007843.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007843.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007843.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200850.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200850.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200850.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200850.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200850.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200850.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200850.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200853.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200853.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200853.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200853.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200853.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200853.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200853.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200854.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200854.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200854.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200854.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200854.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200854.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200854.hg.1 | NM_018448.4 | Refseq |
