Assay Details
Gene Symbol: LOC149703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK055783.1 | - | 1 | 1041 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | R3H domain containing like | R3HDML | dJ881L22.3 | -6.9kb | Anti-sense | Chr. 20 - 44337158 - 44350792 on NCBI Build GRCh38 |
Downstream | HNF4A antisense RNA 1 | HNF4A-AS1 | uc002xlx | 19.5kb | Sense | Chr. 20 - 44377743 - 44395706 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
