Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007850.hg.1 |
ENST00000299218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007850.hg.1 |
ENST00000540047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007850.hg.1 |
ENST00000545606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007850.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007850.hg.1 |
TC12000595.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000539109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000539109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000540525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200861.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200200861.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200200861.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200861.hg.1 |
TC1200012657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200200861.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200861.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200200861.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000535146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000539109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000539109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000539434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000540525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000541058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
ENST00000544619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012657.hg.1 |
NM_018448.4 |
Refseq |