Assay Details
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137605.2 | NP_001131077.1 | 12 - 13 | 1236 | 106 | |
NM_022141.6 | NP_071424.1 | 12 - 13 | 1296 | 106 | ||
XM_005261702.3 | XP_005261759.1 | 12 - 13 | 1023 | 106 | ||
XM_011530302.2 | XP_011528604.1 | 11 - 12 | 3788 | 106 | ||
XM_017028907.1 | XP_016884396.1 | 12 - 13 | 2234 | 106 | ||
XM_017028908.1 | XP_016884397.1 | 12 - 13 | 2254 | 106 | ||
GenBank mRNA | AF237772.1 | - | 12 - 13 | 1090 | 106 | |
AK225929.1 | - | 13 - 14 | 1186 | 106 | ||
BC034406.1 | - | 12 - 13 | 1217 | 5178529 | 106 | |
BX648942.1 | - | 7 - 8 | 4153 | 106 | ||
CA440742.1 | - | 2 - 3 | 106 | |||
CR456480.2 | - | 11 - 12 | 867 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000372.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
