Assay Details
Gene Symbol: CENPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024629.3 | NP_078905.2 | 4 - 5 | 386 | 117 | |
NR_104593.1 | 4 - 5 | 386 | 117 | |||
XM_005263218.4 | XP_005263275.2 | 4 - 5 | 421 | 117 | ||
GenBank mRNA | AF469667.1 | - | 4 - 5 | 389 | 117 | |
AF516710.1 | - | 5 - 6 | 803 | 117 | ||
AK027121.1 | - | 2 - 3 | 196 | 117 | ||
AK292297.1 | - | 4 - 5 | 332 | 117 | ||
BC067802.1 | - | 4 - 5 | 326 | 117 | ||
BC131556.1 | - | 4 - 5 | 386 | 40108113 | 117 | |
BC141854.1 | - | 4 - 5 | 384 | 8860370 | 117 | |
DQ907910.1 | - | 4 - 5 | 316 | 117 | ||
DQ907911.1 | - | 4 - 5 | 316 | 117 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001808.hg
Additional Information:
This assay is in the 5'UTR region of the AK027121.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04024646.hg.1 | TC04001808.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04024646.hg.1 | ENST00000281453.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024646.hg.1 | ENST00000541665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024646.hg.1 | ENST00000541971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024646.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186377.hg.1 | TC0400012620.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
