Assay Details
Gene Symbol: P3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146289.1 | NP_001139761.1 | 5 - 6 | 1199 | 151 | |
NM_001243246.1 | NP_001230175.1 | 5 - 6 | 1199 | 151 | ||
NM_022356.3 | NP_071751.3 | 5 - 6 | 1199 | 151 | ||
XM_005271110.2 | XP_005271167.1 | 4 - 5 | 536 | 151 | ||
GenBank mRNA | AF097432.1 | - | 5 - 6 | 1137 | 151 | |
AK027648.1 | - | 5 - 6 | 1127 | 151 | ||
AK027680.1 | - | 5 - 6 | 1101 | 151 | ||
AK027697.1 | - | 5 - 6 | 1138 | 151 | ||
AK075418.1 | - | 5 - 6 | 1116 | 151 | ||
AK297980.1 | - | 6 - 7 | 1004 | 151 | ||
AK300210.1 | - | 6 - 7 | 1090 | 151 | ||
BC108311.1 | - | 5 - 6 | 1137 | 4521167 | 151 | |
BC143961.1 | - | 5 - 6 | 1115 | 151 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002574.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005271110.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
