Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173075.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173075.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173075.hg.1 |
TC0900010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173075.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173075.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
ENST00000436671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010734.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173073.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173073.hg.1 |
NR_109795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173073.hg.1 |
TC0900010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173073.hg.1 |
ENST00000425666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173073.hg.1 |
NR_109795.1 |
Refseq |