Assay Details
Gene Symbol: FLNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164317.1 | NP_001157789.1 | 31 - 32 | 5435 | 67 | |
NM_001457.3 | NP_001448.2 | 30 - 31 | 5342 | 67 | ||
GenBank mRNA | AB209889.1 | - | 11 - 12 | 2211 | 67 | |
AB371580.1 | - | 31 - 32 | 5415 | 67 | ||
AB371582.1 | - | 30 - 31 | 5320 | 67 | ||
AF042166.1 | - | 30 - 31 | 5308 | 67 | ||
AF043045.1 | - | 30 - 31 | 5342 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000375.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
