Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000480322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000480322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000480322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000617554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
ENST00000617554.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000309180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000480322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000480322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000480322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000617554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
ENST00000617554.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012812.hg.1 |
NM_013308.3 |
Refseq |