Assay Details
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145647.3 | NP_663622.2 | 19 - 20 | 2930 | 116 | |
XM_005251102.1 | XP_005251159.1 | 18 - 19 | 2837 | 116 | ||
XM_005251104.1 | XP_005251161.1 | 18 - 19 | 2693 | 116 | ||
XM_011517378.2 | XP_011515680.1 | 19 - 20 | 3445 | 116 | ||
XM_011517379.2 | XP_011515681.1 | 19 - 20 | 2930 | 116 | ||
XM_017013984.1 | XP_016869473.1 | 18 - 19 | 3052 | 116 | ||
XM_017013985.1 | XP_016869474.1 | 18 - 19 | 2925 | 116 | ||
XM_017013986.1 | XP_016869475.1 | 17 - 18 | 2693 | 116 | ||
XM_017013990.1 | XP_016869479.1 | 17 - 18 | 2594 | 116 | ||
GenBank mRNA | AK025686.1 | - | 12 - 13 | 1626 | 116 | |
AK056434.1 | - | 19 - 20 | 2931 | 116 | ||
BC101724.1 | - | 19 - 20 | 2897 | 8069230 | 116 | |
BC111954.1 | - | 19 - 20 | 2897 | 8327422 | 116 | |
BC143523.1 | - | 18 - 19 | 2781 | 116 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
