Assay Details
Gene Symbol: GLRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002064.2 | NP_002055.1 | 2 - 3 | 540 | 67 | |
GenBank mRNA | AF069668.1 | - | 2 - 3 | 344 | 67 | |
BX106369.1 | - | 2 - 3 | 334 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001608.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing small molecule interconversion
cell redox homeostasis
positive regulation of membrane potential
protein deglutathionylation
positive regulation of sodium ion transmembrane transporter activity
electron transfer activity
glutathione disulfide oxidoreductase activity
protein N-terminus binding
glutathione oxidoreductase activity
nucleobase-containing small molecule interconversion
cell redox homeostasis
positive regulation of membrane potential
protein deglutathionylation
positive regulation of sodium ion transmembrane transporter activity
electron transfer activity
glutathione disulfide oxidoreductase activity
protein N-terminus binding
glutathione oxidoreductase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021960.hg.1 | ENST00000379979.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05021960.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021960.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021960.hg.1 | TC05001608.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05021962.hg.1 | ENST00000379979.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05021962.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021962.hg.1 | TC05001608.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05021961.hg.1 | ENST00000379979.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05021961.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05021961.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021961.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021961.hg.1 | TC05001608.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | TC0500011497.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179695.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179696.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179696.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179696.hg.1 | TC0500011497.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500179696.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179696.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179694.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179694.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179694.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179694.hg.1 | TC0500011497.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500179694.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500179694.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500179694.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
