Assay Details
Gene Symbol: SLC33A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190992.1 | NP_001177921.1 | 6 - 7 | 2145 | 108 | |
GenBank mRNA | BC014416.1 | - | 6 - 7 | 2092 | 4559958 | 108 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001929.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03034576.hg.1 | ENST00000359479.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034576.hg.1 | ENST00000392845.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034576.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034576.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034576.hg.1 | TC03001929.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034573.hg.1 | ENST00000359479.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034573.hg.1 | ENST00000392845.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03034573.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034573.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03034573.hg.1 | TC03001929.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | TC0300012881.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196296.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | NM_004733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | TC0300012881.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | ENST00000359479.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | ENST00000392845.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | ENST00000475842.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | NM_001190992.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196299.hg.1 | NM_004733.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 210406 | - | - |
