Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002085.hg.1 |
ENST00000400384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002085.hg.1 |
ENST00000540640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002085.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002085.hg.1 |
TC18000179.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000586735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000586735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000587823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000587823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000587823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000588916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
TC1800007320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150072.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000400384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000540640.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000586735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000586735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000587823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000587823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000587823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000588916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
ENST00000592595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_001292040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007320.hg.1 |
NM_002747.3 |
Refseq |